missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBR1 Recombinant Rabbit Monoclonal Antibody (24GB3190)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557000
This item is not returnable.
View return policy
Description
CBR1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q.
Specifications
| CBR1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CBR1 | |
| 15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NADP(+)]; 20-beta-hydroxysteroid dehydrogenase; AW261796; carbonyl reductase (NADPH) 1; carbonyl reductase [NADPH] 1; Carbonyl reductase [NADPH] 1 (NADPH-dependent carbonyl reductase 1); carbonyl reductase 1; CBR; Cbr 1; Cbr1; CR; CRN; hCBR1; NADPH-dependent carbonyl reductase 1; prostaglandin 9-ketoreductase; Prostaglandin-E(2) 9-reductase; SDR21C1; Short chain dehydrogenase/reductase family 21C member 1; short chain dehydrogenase/reductase family 21C, member 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29224, 873 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3190 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P16152, P47727 | |
| CBR1 | |
| A synthesized peptide derived from human CBR1 (aa 145-185). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction