missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-86595-25ul
This item is not returnable.
View return policy
Description
CBR1 Polyclonal specifically detects CBR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CBR1 | |
| Polyclonal | |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| P16152 | |
| CBR1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TPFHIQAEVTMKTNFFGTRDVCTELLPLIKPQGRVVNVSSIMSVRALKSCSPELQQKFRSETITEEELVGLMNKFVE | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human CBR1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 15-hydroxyprostaglandin dehydrogenase [NADP+], carbonyl reductase (NADPH) 1, carbonyl reductase (NADPH) 1, EC 1.1.1.18410EC 1.1.1.197,15-hydroxyprostaglandin dehydrogenase, carbonyl reductase [NADPH] 1, carbonyl reductase 1, CBR, CRN, EC 1.1.1.184, EC 1.1.1.189, hCBR1, NADPH-dependent carbonyl reductase 1, Prostaglandin 9-ketoreductase, Prostaglandin-E(2) 9-reductase, SDR21C1, short chain dehydrogenase/reductase family 21C, member 1 | |
| Rabbit | |
| 30 kDa | |
| 25 μL | |
| Stem Cell Markers | |
| 873 | |
| Rat, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction