missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBARA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£247.00 - £460.00
Specifications
| Antigen | CBARA1 |
|---|---|
| Concentration | 0.1mg/mL |
| Dilution | Western Blot 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18422341
|
Novus Biologicals
NBP1-86663-25ul |
25 μL |
£247.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18006874
|
Novus Biologicals
NBP1-86663 |
0.1 mL |
£460.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBARA1 Polyclonal specifically detects CBARA1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CBARA1 | |
| Western Blot 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Allergen Hom s 4, ara CALC, Atopy-related autoantigen CALC, CALCDKFZp564C246, calcium binding atopy-related autoantigen 1, Calcium-binding atopy-related autoantigen 1, MICU1mitochondrial, mitochondrial calcium uptake 1 | |
| MICU1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.1mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| Q9BPX6 | |
| 10367 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LKGKLTIKNFLEFQRKLQHDVLKLEFERHDPVDGRITERQFGGMLLAYSGVQSKKLTAMQRQLKKHFKEGKGLTFQEVENFFTFL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 54 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title