missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBARA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-86663
This item is not returnable.
View return policy
Description
CBARA1 Polyclonal specifically detects CBARA1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CBARA1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Allergen Hom s 4, ara CALC, Atopy-related autoantigen CALC, CALCDKFZp564C246, calcium binding atopy-related autoantigen 1, Calcium-binding atopy-related autoantigen 1, MICU1mitochondrial, mitochondrial calcium uptake 1 | |
| Rabbit | |
| 54 kDa | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.1mg/mL | |
| Western Blot 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q9BPX6 | |
| MICU1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LKGKLTIKNFLEFQRKLQHDVLKLEFERHDPVDGRITERQFGGMLLAYSGVQSKKLTAMQRQLKKHFKEGKGLTFQEVENFFTFL | |
| Affinity Purified | |
| RUO | |
| 10367 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction