missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caveolin 3 Recombinant Rabbit Monoclonal Antibody (23GB3035), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Caveolin 3 |
|---|---|
| Clone | 23GB3035 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088827
|
Invitrogen™
MA553328 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088569
|
Invitrogen™
MA553329 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caveolin 3 Recombinant Monoclonal antibody specifically detects Caveolin 3 in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Caveolin 3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| P51638, P56539 | |
| 29161, 859 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3035 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI385751; CAV; Cav3; Cav-3; caveolin 3; caveolin-3; LGMD1C; LQT9; M-cav; M-caveolin; MGC126100; MGC126101; MGC126129; structural protein; VIP21; VIP-21 | |
| A synthesized peptide derived from Caveolin-3 (1-60AA). | |
| Primary | |
| -20°C | |
| Cav3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title