missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caveolin 3 Recombinant Rabbit Monoclonal Antibody (23GB3035), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553328
This item is not returnable.
View return policy
Description
Caveolin-3 (CAV3) acts as a scaffolding protein within the caveolar membrane. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-3 may also regulate voltage-gated potassium channels. It also plays a role in the sarcolemma repair mechanism of both skeletal muscle and cardiomyocytes that permits rapid resealing of membranes disrupted by mechanical stress. Caveolin-3 also mediates the recruitment of CAVIN2 and CAVIN3 proteins to the caveolae. Mutations in the gene can result in HyperCKmia, rippling muscle disease 2, cardiomyopathy, long QT syndrome 9, sudden infant death syndrome, and myopathy.
Specifications
| Caveolin 3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Cav3 | |
| AI385751; CAV; Cav3; Cav-3; caveolin 3; caveolin-3; LGMD1C; LQT9; M-cav; M-caveolin; MGC126100; MGC126101; MGC126129; structural protein; VIP21; VIP-21 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29161, 859 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3035 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51638, P56539 | |
| Cav3 | |
| A synthesized peptide derived from Caveolin-3 (1-60AA). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction