missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cav3.2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£140.00 - £299.00
Specifications
| Antigen | Cav3.2 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18049925
|
Novus Biologicals
NBP3-15334-20UL |
20 μg |
£140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18307454
|
Novus Biologicals
NBP3-15334-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cav3.2 Polyclonal antibody specifically detects Cav3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Cav3.2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CACNA1HB, calcium channel, voltage-dependent, T type, alpha 1H subunit, calcium channel, voltage-dependent, T type, alpha 1Hb subunit, Cav3.2, ECA6, EIG6, FLJ90484, Low-voltage-activated calcium channel alpha1 3.2 subunit, low-voltage-activated calcium channel alpha13.2 subunit, voltage dependent t-type calcium channel alpha-1H subunit, voltage-dependent T-type calcium channel subunit alpha-1H, voltage-gated calcium channel alpha subunit Cav3.2, voltage-gated calcium channel alpha subunit CavT.2, Voltage-gated calcium channel subunit alpha Cav3.2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1890-2030 of human Cav3.2 (NP_066921.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 8912 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title