missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cav3.2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-15334-100UL
This item is not returnable.
View return policy
Description
Cav3.2 Polyclonal antibody specifically detects Cav3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Cav3.2 | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| CACNA1HB, calcium channel, voltage-dependent, T type, alpha 1H subunit, calcium channel, voltage-dependent, T type, alpha 1Hb subunit, Cav3.2, ECA6, EIG6, FLJ90484, Low-voltage-activated calcium channel alpha1 3.2 subunit, low-voltage-activated calcium channel alpha13.2 subunit, voltage dependent t-type calcium channel alpha-1H subunit, voltage-dependent T-type calcium channel subunit alpha-1H, voltage-gated calcium channel alpha subunit Cav3.2, voltage-gated calcium channel alpha subunit CavT.2, Voltage-gated calcium channel subunit alpha Cav3.2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1890-2030 of human Cav3.2 (NP_066921.2). | |
| 100 μg | |
| Neuroscience, Signal Transduction | |
| 8912 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction