missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CATSPERE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£415.00
Specifications
| Antigen | C1orf101 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CATSPERE Polyclonal specifically detects CATSPERE in Human samples. It is validated for Western Blot.Specifications
| C1orf101 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 257044 | |
| IgG | |
| 91 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q5SY80.2 | |
| C1orf101 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title