missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin H/K/L/V Recombinant Rabbit Monoclonal Antibody (23GB5780), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Cathepsin H/K/L/V |
|---|---|
| Clone | 23GB5780 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089104
![]() |
Invitrogen™
MA553922 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087819
|
Invitrogen™
MA553923 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin H/K/L/V Recombinant Monoclonal antibody specifically detects Cathepsin H/K/L/V in Human, Mouse samples. It is validated for Western BlotSpecifications
| Cathepsin H/K/L/V | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O60911, P06797, P07711, P09668, P43235, P49935, P55097 | |
| 13036, 13038, 13039, 1512, 1513, 1514, 1515 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5780 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ACC4; ACC-4; ACC5; ACC-5; AI323530; AL022844; aleurain; Cat H; Cat K; cathepsin B3; Cathepsin BA; cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; Cathepsin K; cathepsin L2; cathepsin L2, preproprotein; cathepsin O; cathepsin O1; cathepsin O2; Cathepsin U; cathepsin V; Cathepsin X; catK; CATL2; CPSB; CTS02; CTSH; Ctsk; CTSL2; CTSO; CTSO1; CTSO2; CTSU; CTSV; minisatellite 10q detected by probe MMS10; MMS10-Q; Ms10q; N-benzoylarginine-beta-naphthylamide hydrolase; PKND; Pro-cathepsin H; PYCD; UNQ268/PRO305 | |
| A synthesized peptide derived from human Cathepsin L/V/K/H (110-135AA). | |
| Primary | |
| -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title