missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin H/K/L/V Recombinant Rabbit Monoclonal Antibody (23GB5780), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553922
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a lysosomal cysteine proteinase that plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. The encoded protein has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria. This protein, which is a member of the peptidase C1 family, is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
| Cathepsin H/K/L/V | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CTSV | |
| ACC4; ACC-4; ACC5; ACC-5; AI323530; AL022844; aleurain; Cat H; Cat K; cathepsin B3; Cathepsin BA; cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; Cathepsin K; cathepsin L2; cathepsin L2, preproprotein; cathepsin O; cathepsin O1; cathepsin O2; Cathepsin U; cathepsin V; Cathepsin X; catK; CATL2; CPSB; CTS02; CTSH; Ctsk; CTSL2; CTSO; CTSO1; CTSO2; CTSU; CTSV; minisatellite 10q detected by probe MMS10; MMS10-Q; Ms10q; N-benzoylarginine-beta-naphthylamide hydrolase; PKND; Pro-cathepsin H; PYCD; UNQ268/PRO305 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13036, 13038, 13039, 1512, 1513, 1514, 1515 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 23GB5780 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60911, P06797, P07711, P09668, P43235, P49935, P55097 | |
| CTSH, CTSK, CTSL, CTSV | |
| A synthesized peptide derived from human Cathepsin L/V/K/H (110-135AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction