Learn More
Cathepsin G Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-33498
This item is not returnable.
View return policy
Description
Cathepsin G Polyclonal specifically detects Cathepsin G in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cathepsin G | |
| Polyclonal | |
| cathepsin G, CathepsinG, CGCATG, EC 3.4.21, EC 3.4.21.20, MGC23078 | |
| RUO | |
| IgG |
| Immunohistochemistry | |
| Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| 0.1 mL | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |