Learn More
Abcam Cathepsin B Inhibitor Screening Kit (Fluorometric)
Product Code. 32111730 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
Product Code. Quantity Unit Size
32111730 100 Tests 100 tests
1 options
Product Code. 32111730 Supplier Abcam Supplier No. ab185438100Test
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Abcam's Cathepsin B Inhibitor Screening Kit (ab185438) utilizes the ability of Cathepsin B to cleave the synthetic AFC based peptide substrate to release AFC, which can be easily quantified using a fluorometer or fluorescence microplate reader.
Abcam's Cathepsin B Inhibitor Screening Kit (ab185438) utilizes the ability of Cathepsin B to cleave the synthetic AFC based peptide substrate to release AFC, which can be easily quantified using a fluorometer or fluorescence microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests