missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin, beta Mouse, Clone: 6F9, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | Catenin, beta |
---|---|
Clone | 6F9 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15949867
|
EnquireBio
1499-MSM5-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939867
|
EnquireBio
1499-MSM5-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Catenin, beta | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
1499 | |
Recombinant chicken ?-catenin | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
92kDa |
6F9 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001895 | |
CTNNB1 | |
IgG1 κ | |
Protein A/G Purified | |
Cell surface and cytoplasmic | |
HeLa or MCF-7 cells. Breast carcinoma |