missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00
Specifications
| Antigen | Catenin alpha 1 |
|---|---|
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Catenin alpha 1 Polyclonal specifically detects Catenin alpha 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Catenin alpha 1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Core ESC Like Genes, Cytoskeleton Markers, Stem Cell Markers | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| 1495 | |
| Synthetic peptide: EKQDETQTKIKRASQ conjugated to KLH, corresponding to amino acids 873-887 of Human Catenin alpha 1 Antibody. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13 | |
| CTNNA1 | |
| IgG | |
| IgG purified | |
| Catenin alpha 1 recognizes alpha-E-catenin (102 kDa).Staining of the Catenin alpha 1 band in immunoblotting is specifically inhibited with the Catenin alpha 1 immunizing peptide (human, amino acids 873-887). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title