missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-11346-0.025ml
This item is not returnable.
View return policy
Description
Catenin alpha 1 Polyclonal specifically detects Catenin alpha 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Catenin alpha 1 | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13 | |
| Rabbit | |
| IgG purified | |
| RUO | |
| Primary | |
| Catenin alpha 1 recognizes alpha-E-catenin (102 kDa).Staining of the Catenin alpha 1 band in immunoblotting is specifically inhibited with the Catenin alpha 1 immunizing peptide (human, amino acids 873-887). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| CTNNA1 | |
| Synthetic peptide: EKQDETQTKIKRASQ conjugated to KLH, corresponding to amino acids 873-887 of Human Catenin alpha 1 Antibody. | |
| 0.025 mL | |
| Cancer, Cellular Markers, Core ESC Like Genes, Cytoskeleton Markers, Stem Cell Markers | |
| 1495 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction