Learn More
Fisherbrand™ Catalyst Tygon E-Food Two-Stop Microbore Tubing Sets
Product Code. 15551395 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.19 mm
0.25 mm
0.89 mm
1.42 mm
2.06 mm
2.79 mm
Unit Size:
Pack of 12
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
15551395 2.79 mm Pack of 12
15511395 0.19 mm Pack of 12
15521395 0.25 mm Pack of 12
15531395 0.89 mm Pack of 12
15541395 1.42 mm Pack of 12
15408914 2.06 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15551395
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Preconfigured ready-to-use links and assemblies.
  • For Small cartridges
  • Time-saving assembly
  • Four formulations for range of chemical compatibility
  • Use with Catalyst extension coils/tubing
Specifications
Diameter (Metric) Inner 2.79 mm