missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAT3/SLC7A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74144
This item is not returnable.
View return policy
Description
CAT3/SLC7A3 Polyclonal specifically detects CAT3/SLC7A3 in Mouse samples. It is validated for Western Blot.
Specifications
| CAT3/SLC7A3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ATRC3cationic amino acid transporter 3, CAT3, CAT-3Cationic amino acid transporter y+, FLJ14541, MGC20687, solute carrier family 7 (cationic amino acid transporter, y+ system), member 3, Solute carrier family 7 member 3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84889 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P70423 | |
| SLC7A3 | |
| Synthetic peptides corresponding to the middle region of Slc7a3. Immunizing peptide sequence RAWSSAFDNLIGNHISRTLKGTILLKMPHVLAEYPDFFALALVLLLTGLL. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Xenopus: 76%; Chicken: 76%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction