missing translation for 'onlineSavingsMsg'
Learn More

Casticin, TRC

Product Code. 30133786
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133786 50 mg 50mg
30012843 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133786 Supplier Toronto Research Chemicals Supplier No. TRCC18600050MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-91-4
Molecular Formula C19 H18 O8
Molecular Weight (g/mol) 374.34
Synonym 4H-1-Benzopyran-4-one, 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxy-, Casticin (6CI), Flavone, 3',5-dihydroxy-3,4',6,7-tetramethoxy- (7CI,8CI), 3,6,7,4'-Tetra-O-methyl-5,3'-dihydroxyflavone, 3,6,7,4'-Tetra-O-methylquercetagetin, 5,12-Dihydroxy-2,6,7,13-tetramethoxyflavone, 5,3'-Dihydroxy-3,6,7,4'-tetramethoxyflavone, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxy-4H-1-benzopyran-4-one, Casticine, Quercetagetin 3,6,7,4'-tetramethyl ether, Vitexicarpin
IUPAC Name 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxychromen-4-one
SMILES COC1=C(Oc2cc(OC)c(OC)c(O)c2C1=O)c3ccc(OC)c(O)c3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H18O8/c1-23-11-6-5-9(7-10(11)20)17-19(26-4)16(22)14-12(27-17)8-13(24-2)18(25-3)15(14)21/h5-8,20-21H,1-4H3
Formula Weight 374.1002
Chemical Name or Material Casticin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.