Learn More
GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 Cells
GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 Cells contain the human calcium-sensing receptor.
Brand: GeneBLAzer™ K1833
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Gene Symbol: CaSR
- Accession Number: NM_000388
- Population Status: Single Cell Clone Origin
- GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 cells contain the human calcium-sensing receptor (CaSR), (Accession # NM_000388) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 (Cat. No. K1536) cells contain a beta-lactamase reporter gene under control of the NFAT Response Element.
- The GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of Spermine. In addition, GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
Specifications
Liquid Nitrogen | |
GPCRs | |
8 x 106 Cells |
Fluorescent | |
CaSR-Gqo5-NFAT-bla CHO-K1 cells, CaSR-Gqo5-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
Dividing Cells |