missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CaspGLOW™ Fluorescein Active Caspase Staining Kit

Product Code. 15540987
Click to view available options
:
CaspGLOW Fluorescein Active Caspase Staining Kit
This item is not returnable. View return policy

Product Code. 15540987

Brand: Invitrogen™ 88700342

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • FITC-VAD-FMK
  • Wash Buffer
  • Z-VAD-FMK

Contains the reagents to sensitively detect active Caspases in mammalian cells. Detection of the labeled cells can be determined by flow cytometry, fluorescent microscopy or a fluorescent plate reader.

  • Uses the inhibitor specific for all caspases, Z-VAD-FMK, which is directly conjugated to FITC (fluorescein isothiocyanate) for the detection system
  • This reagent is cell permeable, non-toxic and irreversibly binds to the active enzyme
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 Tests
Format Kit
Product Type Staining Kit
For Use With (Application) Flow Cytometry
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.