missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 9/p35/p10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Spezifikation
| Antigen | Caspase 9/p35/p10 |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
16846733
|
Proteintech
10380-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16836733
|
Proteintech
10380-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
10380-1-AP can recognize the pre- and cleaved- caspase 9.
Involved in the activation cascade of caspases responsible for apoptosis execution. Binding of caspase-9 to Apaf-1 leads to activation of the protease which then cleaves and activates caspase-3. Promotes DNA damage-induced apoptosis in a ABL1/c-Abl-dependent manner. Proteolytically cleaves poly(ADP-ribose) polymerase (PARP).Spezifikation
| Caspase 9/p35/p10 | |
| Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| P55211, Q8C3Q9 | |
| 12371, 58918, 842 | |
| Caspase 9/p35/p10 Fusion Protein Ag0404 | |
| Primary | |
| -20°C | |
| CASP9 |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| APAF 3, APAF3, Apoptotic protease Mch 6, CASP 9, CASP9, Caspase 9, CASPASE 9c, Caspase9, Caspase-9, ICE LAP6, ICE like apoptotic protease 6, MCH6 | |
| Casp9 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts