missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 9/p35/p10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10380-1-AP-20UL
This item is not returnable.
View return policy
Description
10380-1-AP can recognize the pre- and cleaved- caspase 9.
Involved in the activation cascade of caspases responsible for apoptosis execution. Binding of caspase-9 to Apaf-1 leads to activation of the protease which then cleaves and activates caspase-3. Promotes DNA damage-induced apoptosis in a ABL1/c-Abl-dependent manner. Proteolytically cleaves poly(ADP-ribose) polymerase (PARP).Specifications
| Caspase 9/p35/p10 | |
| Polyclonal | |
| Unconjugated | |
| CASP9 | |
| APAF 3, APAF3, Apoptotic protease Mch 6, CASP 9, CASP9, Caspase 9, CASPASE 9c, Caspase9, Caspase-9, ICE LAP6, ICE like apoptotic protease 6, MCH6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12371, 58918, 842 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P55211, Q8C3Q9 | |
| Casp9 | |
| Caspase 9/p35/p10 Fusion Protein Ag0404 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction