missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-9 Antibody (LAP6 96-2-22) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
£171.00 - £386.00
Specifications
| Antigen | Caspase-9 |
|---|---|
| Clone | LAP6 96-2-22 |
| Concentration | 30.7 mg/mL |
| Dilution | Western Blot 1 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 8-25 ug/ml. Use reported by customer review, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 8-25 ug/ml, KnockDown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18202350
|
Novus Biologicals
NB500-209SS |
0.05 mg |
£171.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18716643
|
Novus Biologicals
NB500-209 |
0.2 mg |
£386.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-9 Monoclonal specifically detects Caspase-9 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Caspase-9 | |
| 30.7 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Caspases, Cell Biology | |
| Ascites with 0.1% Sodium Azide | |
| APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease | |
| CASP9 | |
| IgG1 κ | |
| Unpurified |
| LAP6 96-2-22 | |
| Western Blot 1 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 8-25 ug/ml. Use reported by customer review, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 8-25 ug/ml, KnockDown Validated | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| P55211 | |
| 842 | |
| Recombinant human Caspase 9 pro-domain protein. [UniProt# P55211] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title