missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-9 Antibody (LAP6 96-2-22) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB500-209SS
This item is not returnable.
View return policy
Description
Caspase-9 Monoclonal specifically detects Caspase-9 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| Caspase-9 | |
| Monoclonal | |
| 30.7 mg/mL | |
| Western Blot 1 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 8-25 ug/ml. Use reported by customer review, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 8-25 ug/ml, KnockDown Validated | |
| P55211 | |
| CASP9 | |
| Recombinant human Caspase 9 pro-domain protein. [UniProt# P55211] | |
| 0.05 mg | |
| Apoptosis, Cancer, Caspases, Cell Biology | |
| 842 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| LAP6 96-2-22 | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction