missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caspase 6 Monoclonal Antibody (C6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551056
This item is not returnable.
View return policy
Description
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
Specifications
| Caspase 6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q3T0P5 | |
| Casp6 | |
| Recombinant Bovine CASP6 protein, Ala194-Asn293 (Accession #Q3T0P5). | |
| 20 μL | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| C6 | |
| Unconjugated | |
| Casp6 | |
| apoptotic protease MCH-2; CASP6; CASP-6; caspase 6; caspase 6, apoptosis-related cysteine peptidase; caspase 6, apoptosis-related cysteine protease; caspase-6; Caspase-6 subunit p11; Caspase-6 subunit p18; mCASP-6; MCH2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 538409 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction