missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-12 Antibody (14F7) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£175.00
Specifications
| Antigen | Caspase-12 |
|---|---|
| Clone | 14F7 |
| Concentration | 2 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Microarray |
| Applications | Western Blot |
Description
Caspase-12 Monoclonal specifically detects Caspase-12 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Caspase-12 | |
| 2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Apoptosis, Caspases | |
| Apoptosis Related Cysteine Protease, CASP 12, casp12, CASP12P1, Caspase 12 pseudogene 1, Caspase12, UNQ9415 | |
| CASP12 | |
| IgG2a | |
| Protein A or G purified | |
| This antibody recognizes the full length and possibly the truncated p20 subunit of Caspase 12. |
| 14F7 | |
| Western Blot 1-2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Microarray | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 100506742 | |
| Recombinant fragment: GPQKICTPSS PSESKRKVEDD EMEVNAGLAH ESHLM LTAPH GLQSSEVQDTL KLCPRDQFCK IKTERAKEIY PVMEKEGRTRL ALIICNKKFDY LFDRDNADTDI LNMQELLENLG YSVVLKENLTA QEMETELMQFA GRPEHQSSDST FLVFMSHGIL EGICGVKHRNK KPDVLHDDTIF KIFNNSNCRSL RNKPKILIMQA CRGRYNGTIWV STNKGIATAD, corresponding to amino acids 95-318 of Mouse Caspase 12. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title