missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-12 Antibody (14F7) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB600-1432-0.025ml
This item is not returnable.
View return policy
Description
Caspase-12 Monoclonal specifically detects Caspase-12 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Caspase-12 | |
| Monoclonal | |
| 2 mg/mL | |
| Western Blot 1-2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Microarray | |
| CASP12 | |
| Recombinant fragment: GPQKICTPSS PSESKRKVEDD EMEVNAGLAH ESHLM LTAPH GLQSSEVQDTL KLCPRDQFCK IKTERAKEIY PVMEKEGRTRL ALIICNKKFDY LFDRDNADTDI LNMQELLENLG YSVVLKENLTA QEMETELMQFA GRPEHQSSDST FLVFMSHGIL EGICGVKHRNK KPDVLHDDTIF KIFNNSNCRSL RNKPKILIMQA CRGRYNGTIWV STNKGIATAD, corresponding to amino acids 95-318 of Mouse Caspase 12. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This antibody recognizes the full length and possibly the truncated p20 subunit of Caspase 12. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot | |
| 14F7 | |
| Unconjugated | |
| Apoptosis Related Cysteine Protease, CASP 12, casp12, CASP12P1, Caspase 12 pseudogene 1, Caspase12, UNQ9415 | |
| Rat | |
| 50 kDa | |
| 0.025 mL | |
| Apoptosis, Caspases | |
| 100506742 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction