missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-10/FLICE2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£157.00 - £234.00
Specifications
| Antigen | Caspase-10/FLICE2 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18007073
|
Novus Biologicals
NB100-56125SS |
0.025 mL |
£157.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18076264
|
Novus Biologicals
NB100-56125 |
0.05 mL |
£234.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-10/FLICE2 Polyclonal specifically detects Caspase-10/FLICE2 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Caspase-10/FLICE2 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer, Caspases | |
| Whole antisera with 0.02% Sodium Azide | |
| 843 | |
| Recombinant full-length human caspase-10 was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Apoptotic protease Mch-4, CASP-10, caspase 10, apoptosis-related cysteine peptidase, caspase 10, apoptosis-related cysteine protease, caspase-10, EC 3.4.22.63, FADD-like ICE2, FAS-associated death domain protein interleukin-1B-converting enzyme 2, FLICE2, ICE-like apoptotic protease 4, interleukin-1B-converting enzyme 2, MCH4ALPS2 | |
| CASP10 | |
| IgG | |
| Unpurified | |
| This polyclonal antisera recognizes pro and active forms of caspase-10. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title