missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-10/FLICE2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56125
This item is not returnable.
View return policy
Description
Caspase-10/FLICE2 Polyclonal specifically detects Caspase-10/FLICE2 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Caspase-10/FLICE2 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Apoptotic protease Mch-4, CASP-10, caspase 10, apoptosis-related cysteine peptidase, caspase 10, apoptosis-related cysteine protease, caspase-10, EC 3.4.22.63, FADD-like ICE2, FAS-associated death domain protein interleukin-1B-converting enzyme 2, FLICE2, ICE-like apoptotic protease 4, interleukin-1B-converting enzyme 2, MCH4ALPS2 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| This polyclonal antisera recognizes pro and active forms of caspase-10. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| CASP10 | |
| Recombinant full-length human caspase-10 was used as immunogen. | |
| 0.05 mL | |
| Apoptosis, Breast Cancer, Cancer, Caspases | |
| 843 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction