missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Caspase-1 Inhibitor I

Product Code. 15496359
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496359 5 mg 5mg
15406369 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496359 Supplier Thermo Scientific Chemicals Supplier No. J64407.#0
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

A potent, specific, and reversible inhibitor of caspases-1, -4 and -5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143313-51-3
Molecular Formula C23H32N4O8
Synonym Ac-YVAD-CHO; Ac-Tyr-Val-Ala-Asp-CHO

Specifications

Quantity 5 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A potent, specific, and reversible inhibitor of caspases-1, -4 and -5
Formula Weight 492.52
Sensitivity Moisture Sensitive
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.