missing translation for 'onlineSavingsMsg'
Learn More

CASK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£155.00

Specifications

Antigen CASK
Concentration 1 mg/mL
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18592191
View Documents
Novus Biologicals
NB300-321-0.025mg
0.025 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

CASK Polyclonal specifically detects CASK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

CASK
Western Blot 1 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Polyclonal
Rabbit
Signal Transduction
PBS, 1 mg/ml BSA with 0.05% Sodium Azide
8573
Synthetic Peptide: C(578) E R D S P S T S R Q S P A N(592)
Primary
Store at -20°C. Avoid freeze/thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
Human
Calcium/calmodulin-dependent serine protein kinase, calcium/calmodulin-dependent serine protein kinase (MAGUK family), calcium/calmodulin-dependent serine protein kinase membrane-associatedguanylate kinase, CAMGUK, CMG, EC 2.7.11, EC 2.7.11.1, FGS4CAGH39, FLJ22219, hCASK, LIN2FLJ31914, MICPCH, peripheral plasma membrane protein CASK, Protein lin-2 homolog, TNRC8, trinucleotide repeat containing 8
CASK
IgG
Affinity Purified
By Western blot, this detects a 112 kDa protein representing CASK from AtT20 cell lysate. Immunohistochemical staining of CASK in mouse brain with this results in staining of the choroid plexus.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.