missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-321-0.025mg
This item is not returnable.
View return policy
Description
CASK Polyclonal specifically detects CASK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CASK | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
| CASK | |
| Synthetic Peptide: C(578) E R D S P S T S R Q S P A N(592) | |
| 0.025 mg | |
| Signal Transduction | |
| 8573 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| Calcium/calmodulin-dependent serine protein kinase, calcium/calmodulin-dependent serine protein kinase (MAGUK family), calcium/calmodulin-dependent serine protein kinase membrane-associatedguanylate kinase, CAMGUK, CMG, EC 2.7.11, EC 2.7.11.1, FGS4CAGH39, FLJ22219, hCASK, LIN2FLJ31914, MICPCH, peripheral plasma membrane protein CASK, Protein lin-2 homolog, TNRC8, trinucleotide repeat containing 8 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| By Western blot, this detects a 112 kDa protein representing CASK from AtT20 cell lysate. Immunohistochemical staining of CASK in mouse brain with this results in staining of the choroid plexus. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction