missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1 gamma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80262
This item is not returnable.
View return policy
Description
Casein Kinase 1 gamma Polyclonal specifically detects Casein Kinase 1 gamma in Mouse samples. It is validated for Western Blot.
Specifications
| Casein Kinase 1 gamma | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| casein kinase 1, gamma 1, casein kinase I isoform gamma-1, CKI-gamma 1, EC 2.7.11, EC 2.7.11.1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 53944 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_775277 | |
| CSNK1G1 | |
| Synthetic peptide directed towards the middle region of mouse CSNK1G1. Peptide sequence CENFPEEMATYLRYVRRLDFFEKPDYEYLRTLFTDLFERKGYTFDYAYDW. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Guinea pig: 100%; Equine: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Human: 92%; Rat: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction