missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Cascade Blue™ Acetyl Azide, Trisodium Salt
Cascade Blue™ acetyl azide, trisodium salt
Brand: Molecular Probes™ C2284
Code : 50
Additional Details : Weight : 0.00525kg
Description
The amine-reactive Cascade Blue™ acetyl azide and its conjugates yield blue-fluorescence (approximate excitation/emission maxima ∼396/410 nm) that can be used as a contrasting color in multicolor applications. When compared with aminocoumarin derivatives, the sulfonated pyrene Cascade Blue™ dye shows less spectral overlap with fluorescein. In addition, Cascade Blue™ dye exhibits high absorptivity, is highly fluorescent and, unlike most dyes, resists quenching upon protein conjugation.Specifications
5 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Cascade Blue™ | |
CASCADE BLUE™ | |
396 |
Acetyl Azide | |
Amine | |
Active Ester, Succinimidyl Ester | |
Classic Dyes | |
410 |
For Research Use Only. Not for use in diagnostic procedures.