Learn More
Ekastu Safety™ CARINA KLEIN DESIGN™ Safety Glasses with Nose Padding
Description
- DIN EN 166 1 - FT
- Maximum wear comfort with unlimited field of view
- Good side protection
- Soft nose pad for greater comfort
- Scratch-proof (super anti-scratch)
- Scratch-proof (super anti-scratch)
- Fog-free (super anti-fog)
- 99.99% UV protection with innovative UV 380 technology
- Four-position adjustable temples
Specifications
Specifications
| Certifications/Compliance | DIN EN 166 1- FT |
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Clear |
| Frame Color | Clear |
| Lens Material | Polycarbonate |
| Frame Material | Polyamide |
| UV Protection | UV Filter |