missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | CARD9 |
|---|---|
| Concentration | 1 mg/ml |
| Dilution | Western Blot 2.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
CARD9 Polyclonal specifically detects CARD9 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| CARD9 | |
| Western Blot 2.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| PBS with 0.02% Sodium Azide | |
| CANDF2, caspase recruitment domain family, member 9, caspase recruitment domain-containing protein 9, hCARD9 | |
| CARD9 | |
| IgG | |
| Affinity Purified | |
| 59 kDa |
| 1 mg/ml | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AF311287 | |
| 64170 | |
| Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human CARD9. The immunogen is located within the last 50 amino acids of CARD9. Amino Acid Squence: DRENTTGSDNTDDTEGS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title