missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76679-0.025mg
This item is not returnable.
View return policy
Description
CARD9 Polyclonal specifically detects CARD9 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CARD9 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CANDF2, caspase recruitment domain family, member 9, caspase recruitment domain-containing protein 9, hCARD9 | |
| Rabbit | |
| 59 kDa | |
| 0.025 mg | |
| Apoptosis | |
| 64170 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 2.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| AF311287 | |
| CARD9 | |
| Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human CARD9. The immunogen is located within the last 50 amino acids of CARD9. Amino Acid Squence: DRENTTGSDNTDDTEGS | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction