missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase E/CPE Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £360.00
Specifications
| Antigen | Carboxypeptidase E/CPE |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652192
|
Novus Biologicals
NBP2-92655-0.02ml |
0.02 mL |
£153.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628172
|
Novus Biologicals
NBP2-92655-0.1ml |
0.1 mL |
£360.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carboxypeptidase E/CPE Polyclonal antibody specifically detects Carboxypeptidase E/CPE in Human, Mouse samples. It is validated for Western BlotSpecifications
| Carboxypeptidase E/CPE | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 1363 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| carboxypeptidase E, Carboxypeptidase H, CPH, EC 3.4.17, EC 3.4.17.10, Enkephalin convertase, Prohormone-processing carboxypeptidase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 257-476 of human CPE (NP_001864.1). PYDETRSGSAHEYSSSPDDAIFQSLARAYSSFNPAMSDPNRPPCRKNDDDSSFVDGTTNGGAWYSVPGGMQDFNYLSSNCFEITVELSCEKFPPEETLKTYWEDNKNSLISYLEQIHRGVKGFVRDLQGNPIANATISVEGIDHDVTSAKDGDYWRLLIPGNYKLTASAPGYLAITKKVAVPYSPAAGVDFELESFSERKEEEKEELMEWWKMMSETLNF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title