missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase E/CPE Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92655-0.02ml
This item is not returnable.
View return policy
Description
Carboxypeptidase E/CPE Polyclonal antibody specifically detects Carboxypeptidase E/CPE in Human, Mouse samples. It is validated for Western Blot
Specifications
| Carboxypeptidase E/CPE | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| carboxypeptidase E, Carboxypeptidase H, CPH, EC 3.4.17, EC 3.4.17.10, Enkephalin convertase, Prohormone-processing carboxypeptidase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 257-476 of human CPE (NP_001864.1). PYDETRSGSAHEYSSSPDDAIFQSLARAYSSFNPAMSDPNRPPCRKNDDDSSFVDGTTNGGAWYSVPGGMQDFNYLSSNCFEITVELSCEKFPPEETLKTYWEDNKNSLISYLEQIHRGVKGFVRDLQGNPIANATISVEGIDHDVTSAKDGDYWRLLIPGNYKLTASAPGYLAITKKVAVPYSPAAGVDFELESFSERKEEEKEELMEWWKMMSETLNF | |
| 0.02 mL | |
| Neuroscience | |
| 1363 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction