Learn More
Carboxypeptidase A6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13604-1-AP-20UL
 View more versions of this product
Product Code. 16801904
  • £135.00 / 20µL
Description
Members of the M14 metallocarboxypeptidase protein family serve many diverse functions and are divided into three subfamilies based on structure, function and amino acid sequence similarity. As a member of the A/B subfamily, CPA6 (Carboxypeptidase A6) is a 437 amino acid secreted, zinc-binding protein that contains a characteristic propetide at the amino terminus,which is cleaved off upon enzyme activation. Although expressed broadly in embryonic tissues, CPA6 is found at highest levels in the adult olfactory bulb and epididymus. CPA6 functions to cleave large C-terminal hydrophobic residues off of proteins and protein substrates and is likely involved in the extracellular processing of neuropeptides. Mutations in the gene encoding CPA6 have been linked to Duane syndrome, a congential eye-movement disorder characterized by the abnormal development of cranial nerve VI (the abducens nerve). There are three isoforms of CPA6 that exist as a result of alternative splicing events.
Specifications
Specifications
Carboxypeptidase A6
Polyclonal
Unconjugated
CPA6
Carboxypeptidase A6, Carboxypeptidase B, CPA6, CPAH
Rabbit
Antigen Affinity Chromatography
RUO
312913, 329093, 57094
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5U901, Q8N4T0
Cpa6
Carboxypeptidase A6 Fusion Protein Ag4466
20 μL
Primary
Rat, Human, Mouse