missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon, Activated, Powder, J.T. Baker™
£284.36
Chemical Identifiers
CAS | 7440-44-0 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.011 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10519190
|
Avantor J.T.Baker
E343-07 |
500 g | Wide Mouth Amber Glass Bottle | ||||||
Chemical Identifiers
7440-44-0 | |
12.011 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
C | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
Specifications
7440-44-0 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
[C] | |
12.011 | |
CHEBI:27594 | |
Powder |
C | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
12.01 | |
Carbon, Activated |