Learn More
Carbenoxolone disodium salt, 97+%
Description
Carbenoxolone is anti-inflammatory. It is often used as a gap junction inhibitor in neuroscience research. Carbenoxolone is also an inhibitor of the steroid dehydrogenase enzymes 11 β-hydroxysteroid dehydrogenase (human) and 3α, 20 β-hydroxysteroid dehydrogenase (bacterial). It is used for the treatment of digestive tract ulcers, especially in the stomach. Used in the treatment of peptic, mainly gastric, ulcers. It is often used as a gap junction inhibitor in neuroscience research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water to 100 mM.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat.
Specifications
Specifications
| Color | White to Cream |
| Quantity | 1 g |
| Merck Index | 14,1793 |
| Solubility Information | Soluble in water to 100 mM. |
| Formula Weight | 614.7 |
| Percent Purity | ≥97% |
| Physical Form | Solid |
| Chemical Name or Material | Carbenoxolone disodium salt |
Safety and Handling
- Carbenoxolone disodium salt
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Carbenoxolone disodium
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.