accessibility menu, dialog, popup

UserName

CAS RN 7421-40-1

NaNa(S)(R)(S)OOOOCH3H3C(R)(R)(S)(S)(R)(S)OH3COOHCH3CH3HHCH3CH3

CAS RN 7421-40-1

IUPAC Name: disodium (2S,4aS,6aS,6bR,8aR,10S,12aS,12bR,14bR)-10-[(3-carboxylatopropanoyl)oxy]-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-2-carboxylate
Synonyms: disodium succinoyl glycyrrhetinate
Molecular Weight (g/mol): 614.73
Molecular Formula: C34H48Na2O7
InChi Key: BQENDLAVTKRQMS-SBBGFIFASA-L
SMILES: [Na+].[Na+].CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C([O-])=O)OC(=O)CCC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results