missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ LC-lambda (mouse) Affinity Matrix

Product Code. 18446498 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
10mL
50mL
5mL
This item is not returnable. View return policy

Product Code. 18446498

Brand: Thermo Scientific™ 194323050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ LC-lambda (mouse) Affinity Matrix contains a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) recognizing the lambda light chain of mouse Ig's.

CaptureSelect LC-lambda (mouse) Affinity Matrix contains a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) recognizing the lambda light chain of mouse Ig's. The ligand is directed towards a unique domain on the constant part of the lambda light chain of immunoglobulins, which enables purification of mouse immunoglobulins and fab fragments without cross reactivity towards bovine immunoglobulins.This makes the CaptureSelect LC-lambda (mouse) Affinity Matrix a unique resin in terms of specificity.

The ligand is coupled on a 70 μm aldehyde-activated agarose bead. This matrix can be used for the purification and isolation of lambda light chain-containing mouse immunoglobulins and fab fragments from complex sources like cell culture supernatants, plasma, and serum in a single step. Due to its non-reactivity to bovine Ig, FCS can be applied in growth media without disturbing purification.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Matrix Agarose-based, aldehyde-activated
Content And Storage Store at 2°C to 8°C.
Particle Size 70 μm
Form Liquid Suspension
Product Line CaptureSelect
Quantity 50 mL
Type Affinity Matrix
Species Mouse
Stationary Phase Anti-LC-lambda
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.