missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ LC-kappa (mur) Affinity Matrix

Product Code. 18466828 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
10mL
50mL
5mL
This item is not returnable. View return policy

Product Code. 18466828

Brand: Thermo Scientific™ 191315005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ LC-kappa (mur) Affinity Matrix enables purification of murine (mouse, rat, and guinea pig) immunoglobulins and Fab fragments without cross reactivity towards bovine immunoglobulins.

CaptureSelect LC-kappa (mur) Affinity Matrix enables purification of murine (mouse, rat, and guinea pig) immunoglobulins and Fab fragments without cross reactivity towards bovine immunoglobulins. The ligand is directed towards a unique domain on the constant part of the kappa light chain of immunoglobulins, which makes CaptureSelect LC-kappa (mur) Affinity Matrix a unique resin in terms of specificity.

This matrix can be used for the purification and isolation of kappa light chain-containing murine immunoglobulins and Fab fragments from complex sources like plasma, serum, and cell culture supernatants in a single step. Due to its non-reactivity with bovine Ig, FCS can be applied in growth media without disturbing purification.

Specifications

Content And Storage Store at 2°C to 8°C.
Particle Size 35 μm
Form Liquid Suspension
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Stationary Phase Anti-LC-kappa (mur)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.