missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ IgM Ligand Leakage ELISA

Product Code. 18446728 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 Assay
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18446728

Brand: Thermo Scientific™ 810289001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ IgM Ligand Leakage ELISA is designed to detect possible leached ligand of IgM purified using Thermo Scientific™ CaptureSelect™ IgM Affinity Matrix.

CaptureSelect IgM Ligand Leakage ELISA is designed to detect possible leached ligand of IgM purified using CaptureSelect IgM Affinity Matrix. The ligand, a 14 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect IgM Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Assay ELISA
For Use With (Equipment) Microplate Reader
Product Line CaptureSelect
Quantity 1 Assay
Type ELISA Kit
Format Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.