missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ IgG4 (Hu) Affinity Matrix

Product Code. 18496738 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
10mL
50mL
5mL
This item is not returnable. View return policy

Product Code. 18496738

Brand: Thermo Scientific™ 2942902005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ IgG4 (Hu) Affinity Matrix has been specifically designed for the purification and isolation of human IgG4 from complex sources such as plasma, serum, or cell culture supernatants.

CaptureSelect IgG4 (Hu) Affinity Matrix has been specifically designed for the purification and isolation of human IgG4 from complex sources such as plasma, serum, or cell culture supernatants. The affinity ligand selectively binds the Fc region of human IgG4, without cross-reactivity to other antibody subclasses and isotypes. The affinity resin enables a robust and efficient purification process with excellent purity obtained in a single step.

Features of this affinity matrix include:

  • Highly selective for human IgG4
  • No cross-reactivity with other antibody subclasses (IgA, IgD IgE, IgM) or antibody subtypes (IgG1-3)
  • No cross reactivity with other species, allowing use of complex growth media containing fetal calf or horse serum
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect IgG4 affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >15 g IgG4/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 0.1 M glycine, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Product Line CaptureSelect
Type Affinity Matrix
Quantity 5 mL
Form Resin Slurry
Particle Size 65 ±10 μm
Binding Capacity >15 g IgG4/L resin
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Ligand Type IgG4 affinity ligand
Matrix Agarose-based, epoxide activated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.