missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CaptureSelect IgA-CH1 (Hu) Affinity Matrix can be used to purify human IgA. It shows no binding to other species IgA or other immunoglobulins.
This product can be used for purification of:
- IgA1 and IgA2 from plasma and serum
- Mono- and dimeric IgA, including secretory IgA
- IgA Fab fragments
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C. |
| Form | Liquid Suspension |
| Product Line | CaptureSelect |
| Quantity | 5 mL |
| Type | Affinity Matrix |
| Stationary Phase | Anti-IgA-CH1 (Hu) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?