missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Human Growth Hormone Affinity Matrix

Product Code. 18456728 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
10mL
50mL
5mL
Product Code. Quantity unitSize
18456728 5 mL 5mL
18446598 10 mL 10mL
18446488 50 mL 50mL
3 options
This item is not returnable. View return policy

Product Code. 18456728

Brand: Thermo Scientific™ 1943160005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Human Growth Hormone (hGH) Affinity Matrix has been specifically designed for the purification of human growth hormone from recombinant sources.

CaptureSelect Human Growth Hormone (hGH) Affinity Matrix has been specifically designed for the purification of human growth hormone from recombinant sources.

Features of this affinity matrix include:

  • Highly selective for recombinant hGH
  • Efficient purification of recombinant hGH with high yields and retained activity
  • Excellent scalability
  • Non-animal-derived

Recombinant hGH, also known as somatotropin or somatropin, is a peptide hormone that stimulates growth, cell reproduction, and regeneration in humans and other animals. The high selectivity and yields obtained using CaptureSelect Human Growth Hormone Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect hGH affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼3 mg/mL resin
  • Elution conditions: 20 mM citric acid, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol

Specifications

Binding Capacity ∼3 mg/mL
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Approved for shipment at Room Temperature and Wet Ice
Particle Size 65 ±10 μm
Form Agarose Bead
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Ligand Type HGH affinity ligand
Stationary Phase HGH
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.